missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-butene, 98%

Catalog No. AAH2723006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2723006 5 g
AAH2723014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2723006 Supplier Thermo Scientific Chemicals Supplier No. H2723006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 563-46-2 | C5H10 | 70.14 g/mol

2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

Solubility
Soluble in ether, ethanol and benzene. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 563-46-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009333
InChI Key MHNNAWXXUZQSNM-UHFFFAOYSA-N
Synonym 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2
PubChem CID 11240
ChEBI CHEBI:77915
IUPAC Name 2-methylbut-1-ene
SMILES CCC(C)=C

Specifications

Melting Point -137°C
Density 0.651
Boiling Point 31°C to 32°C
Flash Point −48°C (−54°F)
Refractive Index 1.374
Quantity 5 g
UN Number UN2459
Solubility Information Soluble in ether,ethanol and benzene. Insoluble in water.
Formula Weight 70.13
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-butene
Show More Show Less
Hazard Category H224-H304
Hazard Statement GHS H Statement
H224-H304
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 2-METHYL-1-BUTENE
EINECSNumber 209-250-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only