Learn More
2-Methyl-1-buten-3-yne, 97%
CAS: 78-80-8 | C5H6 | 66.103 g/mol
Supplier: Thermo Scientific Chemicals L0611114
Description
2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.
Solubility
Not miscible in water.
Notes
Store at 4°C. Incompatible with heavy metal powders, heat. Protect from heat and direct sunlight. Store away from oxidizing agents.
Specifications
| 2-Methyl-1-buten-3-yne | |
| -113°C | |
| 32°C to 33°C | |
| Pungent | |
| 1.418 | |
| 25 g | |
| 969175 | |
| Not miscible in water. | |
| CC(=C)C#C | |
| 66.103 | |
| 66.1 |
| 78-80-8 | |
| 0.694 | |
| −45°C (−49°F) | |
| C5H6 | |
| MFCD00008599 | |
| UN3295 | |
| isopropenylacetylene, 2-methyl-1-buten-3-yne, valylene, 1-buten-3-yne, 2-methyl, 3-methyl-3-buten-1-yne, 2-methylbutenyne, isopropenyl acetylene, acmc-20anrd, 2-methyl-1-butenyne, 2-methyl-l-buten-3-yne | |
| BOFLDKIFLIFLJA-UHFFFAOYSA-N | |
| 2-methylbut-1-en-3-yne | |
| 62323 | |
| 97% |
Chemical Identifiers
| 78-80-8 | |
| 66.103 | |
| BOFLDKIFLIFLJA-UHFFFAOYSA-N | |
| 62323 | |
| CC(=C)C#C |
| C5H6 | |
| MFCD00008599 | |
| isopropenylacetylene, 2-methyl-1-buten-3-yne, valylene, 1-buten-3-yne, 2-methyl, 3-methyl-3-buten-1-yne, 2-methylbutenyne, isopropenyl acetylene, acmc-20anrd, 2-methyl-1-butenyne, 2-methyl-l-buten-3-yne | |
| 2-methylbut-1-en-3-yne |
Safety and Handling
GHS H Statement
H224-H304-H302-H332-H315-H319-H335
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P372-P374-P380-P501c
H224-H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 201-144-9
RTECSNumber : EN0960000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only