accessibility menu, dialog, popup

UserName

CH3CH2HC

CAS RN 78-80-8

IUPAC Name: 2-methylbut-1-en-3-yne
Synonyms: 1-buten-3-yne, 2-methyl-
Molecular Weight (g/mol): 66.1
Molecular Formula: C5H6
InChi Key: BOFLDKIFLIFLJA-UHFFFAOYSA-N
SMILES: CC(=C)C#C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)