missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-heptadiene (cis- and trans- mixture), TCI America™

Catalog No. M03165ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03165ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03165ML Supplier TCI America Supplier No. M03165ML

Chemical Identifiers

CAS 6766-54-7
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00059240
InChI Key LXKRFDPUBWVICN-SNAWJCMRSA-N
PubChem CID 291987
IUPAC Name (5E)-2-methylhepta-1,5-diene
SMILES CC=CCCC(=C)C

Specifications

Color Colorless
Boiling Point 118°C
Quantity 5 mL
UN Number 1993
Formula Weight 110.20
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,5-heptadiene (cis- and trans- mixture)