accessibility menu, dialog, popup

UserName

CH3CH2H3C(E/Z)

CAS RN 6766-54-7

IUPAC Name: 2-methylhepta-1,5-diene
Synonyms: 2-methylhepta-1,5-diene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: LXKRFDPUBWVICN-UHFFFAOYSA-N
SMILES: CC=CCCC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results