missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-naphthoquinone, 98%

CAS: 58-27-5 | C11H8O2 | 172.183 g/mol

$54.21 - $702.59

Chemical Identifiers

CAS 58-27-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00001681
InChI Key MJVAVZPDRWSRRC-UHFFFAOYSA-N
Synonym menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
PubChem CID 4055
ChEBI CHEBI:28869
IUPAC Name 2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=CC=CC=C2C1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1359309
View Documents
Thermo Scientific Chemicals
A1359309
10 g
Each for $54.21
Only null left
 
AAA1359318
View Documents
Thermo Scientific Chemicals
A1359318
50 g
Each for $142.11
Only null left
 
AAA1359330
View Documents
Thermo Scientific Chemicals
A1359330
250 g
Each for $702.59
Only null left
 
Description

Description

2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

Solubility
Soluble in methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

58-27-5
172.183
MJVAVZPDRWSRRC-UHFFFAOYSA-N
4055
2-methylnaphthalene-1,4-dione
C11H8O2
MFCD00001681
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
CHEBI:28869
CC1=CC(=O)C2=CC=CC=C2C1=O

Specifications

58-27-5
Odorless
MFCD00001681
1908453
14,5831
Soluble in methanol
CC1=CC(=O)C2=CC=CC=C2C1=O
172.183
CHEBI:28869
98%
104°C to 107°C
C11H8O2
10 g
Light sensitive
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
MJVAVZPDRWSRRC-UHFFFAOYSA-N
2-methylnaphthalene-1,4-dione
4055
172.18
2-Methyl-1,4-naphthoquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 200-372-6

RTECSNumber : QL9100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only