missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dithiolane, 99%

Catalog No. AAB2337206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2337206 5 g
AAB2337214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2337206 Supplier Thermo Scientific Chemicals Supplier No. B2337206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5616-51-3 | C4H8S2 | 120.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5616-51-3
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD01729666
InChI Key CARJCVDELAMAEJ-UHFFFAOYSA-N
Synonym 1,3-dithiolane, 2-methyl, 2-methyl-1,3-dithiacyclopentane, unii-4o81r0spr0, 1,3-dithiolane,2-methyl, fema no. 3705, acmc-20anre, acetaldehdye ethylene thioacetal
PubChem CID 21828
IUPAC Name 2-methyl-1,3-dithiolane
SMILES CC1SCCS1

Specifications

Density 1.15
Boiling Point 40°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Unpleasant
Quantity 5 g
Formula Weight 120.23
Percent Purity 99%
Chemical Name or Material 2-Methyl-1,3-dithiolane
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 227-038-2
RTECSNumber JP1163000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only