CAS RN 5616-51-3
IUPAC Name:
2-methyl-1,3-dithiolane
Synonyms:
1,3-dithiolane, 2-methyl-
Molecular Weight (g/mol):
120.23
Molecular Formula:
C4H8S2
InChi Key:
CARJCVDELAMAEJ-UHFFFAOYSA-N
SMILES:
CC1SCCS1
1
–
1
of
1
results
1
Promotions Available
2-Methyl-1,3-dithiolane, 99%
2-Methyl-1,3-dithiolane, 99%
CAS: 5616-51-3 Molecular Formula: C4H8S2 Molecular Weight (g/mol): 120.23 MDL Number: MFCD01729666 InChI Key: CARJCVDELAMAEJ-UHFFFAOYSA-N Synonym: 1,3-dithiolane, 2-methyl,2-methyl-1,3-dithiacyclopentane,unii-4o81r0spr0,1,3-dithiolane,2-methyl,fema no. 3705,acmc-20anre,acetaldehdye ethylene thioacetal PubChem CID: 21828 IUPAC Name: 2-methyl-1,3-dithiolane SMILES: CC1SCCS1