missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. M021025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021025G 25 g
M0210500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M021025G Supplier TCI America Supplier No. M021025G

Chemical Identifiers

CAS 497-26-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00003210
InChI Key HTWIZMNMTWYQRN-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-methyl, 2-methyldioxole, 2-methyldioxolane, methyldioxolane, methyl dioxolane, 2-methyl-1,3-dioxacyclopentane, dioxolane, methyl, unii-94w0r6v90a, acetaldehyde ethylene acetal, methyl-1,3-dioxolane
PubChem CID 10342
IUPAC Name 2-methyl-1,3-dioxolane
SMILES CC1OCCO1

Specifications

Color Colorless
Boiling Point 83°C
Quantity 25 g
UN Number 1993
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-dioxolane
RTECSNumber JI3509000
TSCA Yes