accessibility menu, dialog, popup

UserName

CAS RN 497-26-7

IUPAC Name: 2-methyl-1,3-dioxolane
Synonyms: 1,3-dioxolane, 2-methyl-
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: HTWIZMNMTWYQRN-UHFFFAOYSA-N
SMILES: CC1OCCO1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)