missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dioxolane, 98%

Catalog No. AAA1282418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282418 50 g
AAA1282409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1282418 Supplier Thermo Scientific Chemicals Supplier No. A1282418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 497-26-7 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 497-26-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00003210
InChI Key HTWIZMNMTWYQRN-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-methyl, 2-methyldioxole, 2-methyldioxolane, methyldioxolane, methyl dioxolane, 2-methyl-1,3-dioxacyclopentane, dioxolane, methyl, unii-94w0r6v90a, acetaldehyde ethylene acetal, methyl-1,3-dioxolane
PubChem CID 10342
IUPAC Name 2-methyl-1,3-dioxolane
SMILES CC1OCCO1

Specifications

Density 0.982
Boiling Point 80°C to 83°C
Flash Point −2°C (28°F)
Refractive Index 1.398
Quantity 50 g
UN Number UN1993
Beilstein 102520
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-1,3-dioxolane
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-841-4
RTECSNumber JI3509000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only