missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-cyclopentanedione 97.0+%, TCI America™

Catalog No. M09015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09015G 5 g
M090125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09015G Supplier TCI America Supplier No. M09015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765-69-5
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001406
InChI Key HXZILEQYFQYQCE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
PubChem CID 13005
IUPAC Name 2-methylcyclopentane-1,3-dione
SMILES CC1C(=O)CCC1=O

Specifications

Melting Point 214°C
Color White
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,3-cyclopentanedione