missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-cyclohexanedione 98.0+%, TCI America™

Catalog No. M11335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11335G 5 g
M113325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11335G Supplier TCI America Supplier No. M11335G

Chemical Identifiers

CAS 1193-55-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001587
InChI Key VSGJHHIAMHUZKF-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
PubChem CID 70945
IUPAC Name 2-methylcyclohexane-1,3-dione
SMILES CC1C(=O)CCCC1=O

Specifications

Quantity 5 g
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,3-cyclohexanedione