accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 1193-55-1

IUPAC Name: 2-methylcyclohexane-1,3-dione
Synonyms: 2-methylcyclohexane-1,3-dione
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: VSGJHHIAMHUZKF-UHFFFAOYSA-N
SMILES: CC1C(=O)CCCC1=O
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)