missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,2'-dinaphthyl Ketone, TCI America™

Catalog No. M12841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12841G Supplier TCI America Supplier No. M12841G

Chemical Identifiers

CAS 110876-52-3
Molecular Formula C22H16O
Molecular Weight (g/mol) 296.37
MDL Number MFCD01321179
InChI Key GCDFSVMEIZNGPZ-UHFFFAOYSA-N
Synonym 2-Methyl-1-naphthyl 2-Naphthyl Ketone
PubChem CID 22624812
IUPAC Name (2-methylnaphthalen-1-yl)(naphthalen-2-yl)methanone
SMILES CC1=CC=C2C=CC=CC2=C1C(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 140°C
Color Yellow
Quantity 1 g
Formula Weight 296.37
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,2′-dinaphthyl Ketone