missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxytropone 98.0+%, TCI America™

Numéro de catalogue. M25761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M25761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M25761G Fournisseur TCI America Code fournisseur M25761G
Il en reste null

Identifiants chimiques

CAS 2161-40-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD01734435
InChI Key IQYJSKNHDZZDGA-UHFFFAOYSA-N
Synonym 2-Methoxy-2,4,6-cycloheptatrien-1-one
PubChem CID 16546
IUPAC Name 2-methoxycyclohepta-2,4,6-trien-1-one
SMILES COC1=CC=CC=CC1=O

Spécifications

Melting Point 132°C
Color White-Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxytropone
EINECSNumber (3)-2407
RTECSNumber GU4606000
TSCA No