accessibility menu, dialog, popup

UserName

H3COO

CAS RN 2161-40-2

IUPAC Name: 2-methoxycyclohepta-2,4,6-trien-1-one
Synonyms: 2-methoxytropolone
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: IQYJSKNHDZZDGA-UHFFFAOYSA-N
SMILES: COC1=CC=CC=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results