missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiophene 98.0+%, TCI America™

Supplier:  TCI America M10305G

 View more versions of this product

Catalog No. M10305G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methoxythiophene
Yellow
C5H6OS
5 g
thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
COC1=CC=CS1
114.16
114.16
Liquid
16839-97-7
154°C
MFCD00014526
1993
OKEHURCMYKPVFW-UHFFFAOYSA-N
2-methoxythiophene
85610
≥98.0% (GC)

Chemical Identifiers

16839-97-7
114.16
OKEHURCMYKPVFW-UHFFFAOYSA-N
85610
COC1=CC=CS1
C5H6OS
MFCD00014526
thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
2-methoxythiophene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions