CAS RN 16839-97-7
CAS RN 16839-97-7
IUPAC Name:
2-methoxythiophene
Synonyms:
p-methoxythiophene
Molecular Weight (g/mol):
114.16
Molecular Formula:
C5H6OS
InChi Key:
OKEHURCMYKPVFW-UHFFFAOYSA-N
SMILES:
COC1=CC=CS1
1
–
1
of
1
results
1
2-Methoxythiophene 98.0+%, TCI America™
CAS: 16839-97-7 Molecular Formula: C5H6OS Molecular Weight (g/mol): 114.16 MDL Number: MFCD00014526 InChI Key: OKEHURCMYKPVFW-UHFFFAOYSA-N Synonym: thiophene, 2-methoxy,2-methoxy thiophene,2-methoxy-thiophene,2-methyloxy thiophene,pubchem12545,acmc-1bwzz,2-methoxythiophene,ksc183i2f PubChem CID: 85610 IUPAC Name: 2-methoxythiophene SMILES: COC1=CC=CS1