missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxythiophene 98.0+%, TCI America™
Supplier: TCI America M103025G
Specifications
| 2-Methoxythiophene | |
| Yellow | |
| C5H6OS | |
| 25 g | |
| thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f | |
| COC1=CC=CS1 | |
| 114.16 | |
| 114.16 | |
| Liquid |
| 16839-97-7 | |
| 154°C | |
| MFCD00014526 | |
| 1993 | |
| OKEHURCMYKPVFW-UHFFFAOYSA-N | |
| 2-methoxythiophene | |
| 85610 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 16839-97-7 | |
| 114.16 | |
| OKEHURCMYKPVFW-UHFFFAOYSA-N | |
| 85610 | |
| COC1=CC=CS1 |
| C5H6OS | |
| MFCD00014526 | |
| thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f | |
| 2-methoxythiophene |
Safety and Handling
TSCA : No