missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiazole 98.0+%, TCI America™

Catalog No. M17535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17535G 5 g
M175325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17535G Supplier TCI America Supplier No. M17535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14542-13-3
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD01631143
InChI Key MJJRDTKNLLMJDJ-UHFFFAOYSA-N
Synonym 2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
PubChem CID 575451
IUPAC Name 2-methoxy-1,3-thiazole
SMILES COC1=NC=CS1

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
UN Number 1993
Formula Weight 115.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxythiazole