accessibility menu, dialog, popup

UserName

CAS RN 14542-13-3

IUPAC Name: 2-methoxy-1,3-thiazole
Synonyms: 2-methoxy-1,3-thiazole
Molecular Weight (g/mol): 115.15
Molecular Formula: C4H5NOS
InChi Key: MJJRDTKNLLMJDJ-UHFFFAOYSA-N
SMILES: COC1=NC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results