missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyrazine 98.0+%, TCI America™

Catalog No. M07985ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07985ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M07985ML Supplier TCI America Supplier No. M07985ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3149-28-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00006126
InChI Key WKSXRWSOSLGSTN-UHFFFAOYSA-N
Synonym pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine
PubChem CID 18467
IUPAC Name 2-methoxypyrazine
SMILES COC1=NC=CN=C1

Specifications

Color Yellow
Boiling Point 68°C
Quantity 5 mL
UN Number 1993
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxypyrazine