accessibility menu, dialog, popup

UserName

H3CONN

CAS RN 3149-28-8

IUPAC Name: 2-methoxypyrazine
Synonyms: 2-methoxypyrazine
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: WKSXRWSOSLGSTN-UHFFFAOYSA-N
SMILES: COC1=CN=CC=N1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)