missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene 95.0+%, TCI America™

Catalog No. p-7131320
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
I030325ML 25 mL
I0303100ML 100 mL
I0303500ML 500 mL
3 options

Catalog No. I030325ML

Supplier: TCI America I030325ML

Only null left

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

Color Colorless
Boiling Point 36°C
Quantity 25 mL
UN Number 1993
Formula Weight 72.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxypropene
RTECSNumber UD0800000
TSCA Yes
Recommended Storage Refrigerator