accessibility menu, dialog, popup

UserName

H3COCH3H2C

CAS RN 116-11-0

IUPAC Name: 2-methoxyprop-1-ene
Synonyms: 2-methoxypropene
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: YOWQWFMSQCOSBA-UHFFFAOYSA-N
SMILES: COC(C)=C
Boiling Point 
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)