missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene 95.0+%, TCI America™

Catalog No. I030325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030325ML 25 mL
I0303100ML 100 mL
I0303500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I030325ML Supplier TCI America Supplier No. I030325ML

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

Color Colorless
Boiling Point 36°C
Quantity 25 mL
UN Number 1993
Formula Weight 72.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxypropene
RTECSNumber UD0800000
TSCA Yes
Recommended Storage Refrigerator