missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetone 98.0+%, TCI America™

Catalog No. M079425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M079425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M079425ML Supplier TCI America Supplier No. M079425ML

Chemical Identifiers

CAS 5211-62-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00008770
InChI Key GMBFNZCPZFVKAT-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetone, 1-2-methoxyphenyl propan-2-one, 1-2-methoxyphenyl acetone, o-anisylacetone, o-methoxy phenyl acetone, 2-acetonylanisole, o-methoxyphenylaceton, 2-propanone, 1-2-methoxyphenyl, 2-methoxyphenyl acetone, pubchem23387
PubChem CID 78887
IUPAC Name 1-(2-methoxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1OC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenylacetone
EINECSNumber (9)-0051
TSCA No