missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetone, 97%

Catalog No. p-7027607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2449503 1 g
AAB2449506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2449503 Supplier Thermo Scientific Chemicals Supplier No. B2449503
Only null left

CAS: 5211-62-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5211-62-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00008770
InChI Key GMBFNZCPZFVKAT-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetone, 1-2-methoxyphenyl propan-2-one, 1-2-methoxyphenyl acetone, o-anisylacetone, o-methoxy phenyl acetone, 2-acetonylanisole, o-methoxyphenylaceton, 2-propanone, 1-2-methoxyphenyl, 2-methoxyphenyl acetone, pubchem23387
PubChem CID 78887
IUPAC Name 1-(2-methoxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1OC

Specifications

Density 1.062
Boiling Point 128°C to 131°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.525
Quantity 1 g
Beilstein 2502308
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyphenylacetone
EINECSNumber 226-008-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only