missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyphenylacetic Acid 98.0+%, TCI America™
$54.06 - $54.06
Chemical Identifiers
| CAS | 93-25-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00004321 |
| InChI Key | IVEWTCACRDEAOB-UHFFFAOYSA-N |
| Synonym | 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole |
| PubChem CID | 7134 |
| IUPAC Name | 2-(2-methoxyphenyl)acetic acid |
| SMILES | COC1=CC=CC=C1CC(O)=O |
Chemical Identifiers
| 93-25-4 | |
| 166.18 | |
| IVEWTCACRDEAOB-UHFFFAOYSA-N | |
| 7134 | |
| COC1=CC=CC=C1CC(O)=O |
| C9H10O3 | |
| MFCD00004321 | |
| 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole | |
| 2-(2-methoxyphenyl)acetic acid |
Specifications
| 93-25-4 | |
| White-Yellow | |
| MFCD00004321 | |
| 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole | |
| COC1=CC=CC=C1CC(O)=O | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
| 124°C | |
| C9H10O3 | |
| 25 g | |
| IVEWTCACRDEAOB-UHFFFAOYSA-N | |
| 2-(2-methoxyphenyl)acetic acid | |
| 7134 | |
| ≥98.0% (T) | |
| 2-Methoxyphenylacetic Acid |
Safety and Handling
TSCA : Yes