Learn More
2-Methoxyphenylacetic acid, 99%
CAS: 93-25-4 | C9H10O3 | 166.18 g/mol
Supplier: Thermo Scientific Chemicals A1521114
Description
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 2-Methoxyphenylacetic acid | |
| 122°C to 125°C | |
| MFCD00004321 | |
| 2047573 | |
| Soluble in water. Also soluble in methanol,ether,ethanol and acetone. | |
| COC1=CC=CC=C1CC(O)=O | |
| 166.18 | |
| 166.18 |
| 93-25-4 | |
| C9H10O3 | |
| 25 g | |
| 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole | |
| IVEWTCACRDEAOB-UHFFFAOYSA-N | |
| 2-(2-methoxyphenyl)acetic acid | |
| 7134 | |
| 99% |
Chemical Identifiers
| 93-25-4 | |
| 166.18 | |
| IVEWTCACRDEAOB-UHFFFAOYSA-N | |
| 7134 | |
| COC1=CC=CC=C1CC(O)=O |
| C9H10O3 | |
| MFCD00004321 | |
| 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole | |
| 2-(2-methoxyphenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-231-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only