missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetic acid, 99%

Catalog No. AAA1521118
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1521118 50 g
AAA1521109 10 g
AAA1521114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1521118 Supplier Thermo Scientific Chemicals Supplier No. A1521118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-25-4 | C9H10O3 | 166.18 g/mol

2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 93-25-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004321
InChI Key IVEWTCACRDEAOB-UHFFFAOYSA-N
Synonym 2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
PubChem CID 7134
IUPAC Name 2-(2-methoxyphenyl)acetic acid
SMILES COC1=CC=CC=C1CC(O)=O

Specifications

Melting Point 122°C to 125°C
Quantity 50 g
Beilstein 2047573
Solubility Information Soluble in water. Also soluble in methanol,ether,ethanol and acetone.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2-Methoxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-231-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only