missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenoxyacetic acid, 98+%

Catalog No. AAB2067214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2067214 25 g
AAB2067206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2067214 Supplier Thermo Scientific Chemicals Supplier No. B2067214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1878-85-9 | C9H10O4 | 182.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-85-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00014352
InChI Key IHONYPFTXGQWAX-UHFFFAOYSA-N
Synonym 2-methoxyphenoxy acetic acid, 2-2-methoxyphenoxy acetic acid, 2-methoxyphenoxyacetic acid, guaiacoxyacetic acid, acetic acid, 2-methoxyphenoxy, o-methoxyphenoxy acetic acid, unii-7xvz1x365a, acide o-methoxyphenoxyacetique french, o-methoxy phenoxyacetic acid, acetic acid, o-methoxyphenoxy
PubChem CID 15882
IUPAC Name 2-(2-methoxyphenoxy)acetic acid
SMILES COC1=CC=CC=C1OCC(=O)O

Specifications

Melting Point 118°C to 122°C
Quantity 25 g
Beilstein 1874501
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyphenoxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-526-3
RTECSNumber AI8930700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only