missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenethylamine, 98%

Catalog No. p-4229283
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182170050 5 g Glass bottle
AC182170250 25 g Glass bottle
AC182171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC182170050 Supplier Thermo Scientific Chemicals Supplier No. 182170050

CAS: 2045-79-6 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2045-79-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00008186
InChI Key WSWPCNMLEVZGSM-UHFFFAOYSA-N
Synonym 2-methoxyphenethylamine, 2-2-methoxyphenyl ethylamine, 2-2-methoxyphenyl ethanamine, 2-2-methoxy-phenyl-ethylamine, 2-2-methoxyphenyl ethan-1-amine, benzeneethanamine, 2-methoxy, o-methoxyphenethylamine, pubchem23973, acmc-209fah, ortho-methoxyphenethylamine
PubChem CID 74896
IUPAC Name 2-(2-methoxyphenyl)ethanamine
SMILES COC1=CC=CC=C1CCN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.0300g/mL
Boiling Point 236°C to 237°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.542
Linear Formula CH3OC6H4CH2CH2NH2
Quantity 5 g
Beilstein 13,II,352
Specific Gravity 1.03
Formula Weight 151.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 218-066-6

RUO – Research Use Only