missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynaphthalene, 98%

Catalog No. p-7023767
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1765030 250 g
AAA176500B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1765030 Supplier Thermo Scientific Chemicals Supplier No. A1765030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-04-9 | C11H10O | 158.2 g/mol

2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

Solubility
Soluble in benzene, carbon disulfide, diethyl ether. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 93-04-9
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004061
InChI Key LUZDYPLAQQGJEA-UHFFFAOYSA-N
Synonym naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether
PubChem CID 7119
IUPAC Name 2-methoxynaphthalene
SMILES COC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 70°C to 75°C
Boiling Point 272°C to 274°C
Flash Point ca 139°C (282°F)
Quantity 250 g
Beilstein 1859408
Merck Index 14,5997
Solubility Information Soluble in benzene,carbon disulfide,diethyl ether. Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material 2-Methoxynaphthalene
EINECSNumber 202-213-6
RTECSNumber QJ9468750
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only