missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)benzeneboronic acid, 97%

Catalog No. AAH52858MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52858MD 250 mg
AAH5285803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52858MD Supplier Thermo Scientific Chemicals Supplier No. H52858MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126617-98-9 | C8H11BO3 | 165.983 g/mol

2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 126617-98-9
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD03412110
InChI Key JHPPAFXQJZJGEP-UHFFFAOYSA-N
Synonym 2-methoxymethylphenylboronic acid, 2-methoxymethylbenzeneboronic acid, 2-methoxymethyl phenyl boronic acid, 2-methoxymethyl phenylboronic acid, 2-methoxymethyl benzeneboronic acid, 2-methoxymethylphebylboronic acid, boronic acid, 2-methoxymethyl phenyl, pubchem7997
PubChem CID 3759408
IUPAC Name [2-(methoxymethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1COC)(O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 165.99
Percent Purity 97%
Chemical Name or Material 2-(Methoxymethyl)benzeneboronic acid
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only