missing translation for 'onlineSavingsMsg'
Learn More

(2-Methoxyethyl)thiourea 98.0+%, TCI America™

Catalog No. m25971g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25971G Supplier TCI America Supplier No. M25971G
Only null left

Chemical Identifiers

CAS 102353-42-4
Molecular Formula C4H10N2OS
Molecular Weight (g/mol) 134.20
MDL Number MFCD00060469
InChI Key XLJXJKHWLMYXBE-UHFFFAOYSA-N
PubChem CID 1810725
IUPAC Name (2-methoxyethyl)thiourea
SMILES COCCNC(N)=S

Specifications

Melting Point 76°C
Color White
Quantity 1 g
Formula Weight 134.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2-Methoxyethyl)thiourea