missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7136706
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T130325G 25 g
T1303100G 100 g
T1303500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T130325G Supplier TCI America Supplier No. T130325G

Chemical Identifiers

CAS 17178-10-8
Molecular Formula C10H14O4S
Molecular Weight (g/mol) 230.28
MDL Number MFCD00025910
InChI Key TZXJJSAQSRHKCZ-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid 2-Methoxyethyl Ester
PubChem CID 86977
IUPAC Name 2-methoxyethyl 4-methylbenzene-1-sulfonate
SMILES COCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 10°C
Color Colorless
Boiling Point 141°C
Quantity 25 g
Formula Weight 230.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl p-Toluenesulfonate
EINECSNumber (3)-1890
TSCA Yes