missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Chloroformate 93.0+%, TCI America™

Catalog No. C09285ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09285ML Supplier TCI America Supplier No. C09285ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-12-6
Molecular Formula C4H7ClO3
Molecular Weight (g/mol) 138.547
MDL Number MFCD00058932
InChI Key NDYYWMXJZWHRLZ-UHFFFAOYSA-N
Synonym 2-methoxyethyl chloroformate, chloroformic acid 2-methoxyethyl ester, 2-methoxyethylchloroformate, chloro 2-methoxyethoxy methanone, carbonochloridic acid, 2-methoxyethyl ester, 2-methoxyethylchlorocarbonat, pubchem12774, methoxyethyl chloroformate, acmc-1awq9, 2-methoxy-ethyl chloroformate
PubChem CID 69400
IUPAC Name 2-methoxyethyl carbonochloridate
SMILES COCCOC(=O)Cl

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 mL
UN Number 2742
Formula Weight 138.55
Percent Purity ≥93.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Chloroformate
EINECSNumber (2)-1708
TSCA No
Recommended Storage Refrigerator