accessibility menu, dialog, popup

UserName

CAS RN 628-12-6

IUPAC Name: 2-methoxyethyl carbonochloridate
Synonyms: 2-methoxyethyl carbonochloridate
Molecular Weight (g/mol): 138.55
Molecular Formula: C4H7ClO3
InChi Key: NDYYWMXJZWHRLZ-UHFFFAOYSA-N
SMILES: COCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results