missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Acetate 98.0+%, TCI America™

Catalog No. p-7131996
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M011325ML 25 mL
M0113500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M011325ML Supplier TCI America Supplier No. M011325ML

Chemical Identifiers

CAS 110-49-6
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00008719
InChI Key XLLIQLLCWZCATF-UHFFFAOYSA-N
Synonym methyl glycol acetate, mecsac, acetic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, acetate, methylglykolacetat, ethylene glycol monomethyl ether acetate, methyl cellosolve acetate, acetyl methyl cellosolve, 2-methoxyethanol acetate, methyl glycol monoacetate
PubChem CID 8054
IUPAC Name 2-methoxyethyl acetate
SMILES COCCOC(C)=O

Specifications

Melting Point -65°C
Color Colorless
Boiling Point 143°C
Quantity 25 mL
UN Number 1189
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Acetate
EINECSNumber (2)-0740
RTECSNumber KL5950000
TSCA Yes