accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 110-49-6

IUPAC Name: 2-methoxyethyl acetate
Synonyms: 2-methoxyethyl acetate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: XLLIQLLCWZCATF-UHFFFAOYSA-N
SMILES: COCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results