missing translation for 'onlineSavingsMsg'
Learn More

2-Méthoxyéthyle acétate 98,0+%, TCI America™

Numéro de catalogue. M0113500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M0113500ML 500 mL
M011325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M0113500ML Fournisseur TCI America Code fournisseur M0113500ML

Identifiants chimiques

CAS 110-49-6
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00008719
InChI Key XLLIQLLCWZCATF-UHFFFAOYSA-N
Synonym methyl glycol acetate, mecsac, acetic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, acetate, methylglykolacetat, ethylene glycol monomethyl ether acetate, methyl cellosolve acetate, acetyl methyl cellosolve, 2-methoxyethanol acetate, methyl glycol monoacetate
PubChem CID 8054
IUPAC Name 2-méthoxyéthyle acétate
SMILES COCCOC(C)=O

Spécifications

Melting Point -65°C
Color Incolore
Boiling Point 143°C
Quantity 500 mL
UN Number 1189
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material 2-Methoxyethyl Acetate
EINECSNumber (2)-0740
RTECSNumber KL5950000
TSCA Yes