missing translation for 'onlineSavingsMsg'
Learn More

(2-Methoxyethoxymethyl)triethylammonium Chloride 98.0+%, TCI America™

Catalog No. m06815g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06815G Supplier TCI America Supplier No. M06815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60043-43-8
Molecular Formula C10H24ClNO2
Molecular Weight (g/mol) 225.757
MDL Number MFCD00059973
InChI Key SOEVDEHHMZNQOW-UHFFFAOYSA-M
Synonym Triethyl(2-methoxyethoxymethyl)ammonium Chloride
PubChem CID 13966064
IUPAC Name triethyl(2-methoxyethoxymethyl)azanium;chloride
SMILES CC[N+](CC)(CC)COCCOC.[Cl-]

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 5 g
Formula Weight 225.76
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Methoxyethoxymethyl)triethylammonium Chloride