accessibility menu, dialog, popup

UserName

CH3NCH3H3COOH3CCl

CAS RN 60043-43-8

IUPAC Name: triethyl[(2-methoxyethoxy)methyl]azanium chloride
Synonyms: triethyl[(2-methoxyethoxy)methyl]azanium chloride
Molecular Weight (g/mol): 225.76
Molecular Formula: C10H24ClNO2
InChi Key: SOEVDEHHMZNQOW-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+](CC)(CC)COCCOC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results