missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ACS Reagent (Ethylene Glycol Monomethyl Ether), Reagents

Catalog No. 01337891
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337891 1 L
01337890 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337891 Supplier Labchem Inc Supplier No. C12485201C
Only null left

2-Methoxyethanol, ACS Reagent (Ethylene Glycol Monomethyl Ether), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym Ethylene Glycol Monomethyl Ether, Methyl Cellosolve<superscriptT><superscriptM>
IUPAC Name 2-methoxyethan-1-ol
SMILES COCCO

Specifications

Melting Point -85°C
Color Colorless
Boiling Point 123°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.035
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol, ACS Reagent (Ethylene Glycol Monomethyl Ether)