missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018803
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018803 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018803 Supplier Merck Emd Millipore Supplier No. 889075MLF

2-Methoxyethanol is an organic compound, which is commonly used as a solvent for industrial and consumer products.

Featured Industries: Environmental

Specifications

Ignition Point 548°F
CAS 109-86-4
Vapor Pressure 6.17 mmHg (20°C)
Vapor Density 2.62 (vs. air)
Density 0.965 g/mL (at 25°C)
Flash Point 40°C
Physical Form Neat
Linear Formula CH3 OCH2 CH2 OH
MDL Number MFCD00002867
Melting Point -85°C
Boiling Point 124°C to 125°C
Synonym Ethylene glycol monomethyl ether; Methyl glycol
Type Analytical Standard
Quantity 5 mL
Formula Weight 76.09
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less