missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxycarbonyl)benzenesulfonamide, 98%

Catalog No. aaa1355322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1355322 100 g
AAA1355306 5 g
AAA1355314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1355322 Supplier Thermo Scientific Chemicals Supplier No. A1355322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57683-71-3 | C8H9NO4S | 215.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57683-71-3
Molecular Formula C8H9NO4S
Molecular Weight (g/mol) 215.22
MDL Number MFCD00009808
InChI Key VSOOBQALJVLTBH-UHFFFAOYSA-N
Synonym methyl 2-aminosulfonyl benzoate, 2-carbomethoxybenzenesulfonamide, 2-sulfamoylbenzoic acid methyl ester, 2-methoxycarbonyl benzene sulfonamide, methyl o-sulphamoylbenzoate, benzoic acid, 2-aminosulfonyl-, methyl ester, benzoic acid, 2-sulfamoyl-, methyl ester, 2-methoxycarbonylphenylsulfonamide, 2-methoxycarbonylbenzenesulfonamide, 2-methoxycarbonyl benzenesulphonamide
PubChem CID 42546
ChEBI CHEBI:83512
IUPAC Name methyl 2-sulfamoylbenzoate
SMILES COC(=O)C1=CC=CC=C1S(N)(=O)=O

Specifications

Melting Point 123°C to 125°C
Quantity 100 g
Formula Weight 215.23
Percent Purity 98%
Chemical Name or Material 2-(Methoxycarbonyl)benzenesulfonamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 260-903-2
RTECSNumber DH6830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only