Learn More
2-(Methoxycarbonyl)benzenesulfonamide, 98%
CAS: 57683-71-3 | C8H9NO4S | 215.22 g/mol
Supplier: Thermo Scientific Chemicals A1355314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(Methoxycarbonyl)benzenesulfonamide | |
| 123°C to 125°C | |
| MFCD00009808 | |
| methyl 2-aminosulfonyl benzoate, 2-carbomethoxybenzenesulfonamide, 2-sulfamoylbenzoic acid methyl ester, 2-methoxycarbonyl benzene sulfonamide, methyl o-sulphamoylbenzoate, benzoic acid, 2-aminosulfonyl-, methyl ester, benzoic acid, 2-sulfamoyl-, methyl ester, 2-methoxycarbonylphenylsulfonamide, 2-methoxycarbonylbenzenesulfonamide, 2-methoxycarbonyl benzenesulphonamide | |
| COC(=O)C1=CC=CC=C1S(N)(=O)=O | |
| 215.22 | |
| CHEBI:83512 | |
| 98% |
| 57683-71-3 | |
| C8H9NO4S | |
| 25 g | |
| VSOOBQALJVLTBH-UHFFFAOYSA-N | |
| methyl 2-sulfamoylbenzoate | |
| 42546 | |
| 215.23 |
Chemical Identifiers
| 57683-71-3 | |
| 215.22 | |
| VSOOBQALJVLTBH-UHFFFAOYSA-N | |
| 42546 | |
| methyl 2-sulfamoylbenzoate |
| C8H9NO4S | |
| MFCD00009808 | |
| methyl 2-aminosulfonyl benzoate, 2-carbomethoxybenzenesulfonamide, 2-sulfamoylbenzoic acid methyl ester, 2-methoxycarbonyl benzene sulfonamide, methyl o-sulphamoylbenzoate, benzoic acid, 2-aminosulfonyl-, methyl ester, benzoic acid, 2-sulfamoyl-, methyl ester, 2-methoxycarbonylphenylsulfonamide, 2-methoxycarbonylbenzenesulfonamide, 2-methoxycarbonyl benzenesulphonamide | |
| CHEBI:83512 | |
| COC(=O)C1=CC=CC=C1S(N)(=O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 260-903-2
RTECSNumber : DH6830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only