missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzylamine 97.0+%, TCI America™

Catalog No. p-7132414
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08695ML 5 mL
M086925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08695ML Supplier TCI America Supplier No. M08695ML

Chemical Identifiers

CAS 6850-57-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008110
InChI Key PXJACNDVRNAFHD-UHFFFAOYSA-N
Synonym 2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
PubChem CID 81292
IUPAC Name (2-methoxyphenyl)methanamine
SMILES COC1=CC=CC=C1CN

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 mL
UN Number 2735
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzylamine